(4-butoxyphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

323934-1G 323934-500MG 323934-5G
77469-300EA 415.09 USD
77469-300 77481-976 77492-378
(4-butoxyphenyl)(isopropyl)sulfane ≥95%
(4-butoxyphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.362 g/mol
Storage Temperature: Ambient
CAS Number: 3002431-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR