1-(3-Ethylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

323913-1G 323913-500MG 323913-5G
77469-296EA 397.58 USD
77469-296 77481-972 77492-374
1-(3-Ethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Ethylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.297 g/mol
Storage Temperature: Ambient
CAS Number: 1526357-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR