(2-Bromo-6-fluoro-3-methoxyphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

323856-1G 323856-500MG 323856-5G
77469-288EA 415.09 USD
77469-288 77481-964 77492-366
(2-Bromo-6-fluoro-3-methoxyphenyl)(cyclohexyl)methanol ≥95%
(2-Bromo-6-fluoro-3-methoxyphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₈BrFO₂
Storage Temperature: Ambient
CAS Number: 3002421-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR