(2-Bromo-6-fluoro-3-methoxyphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

323851-1G 323851-500MG 323851-5G
77469-280EA 415.09 USD
77469-280 77481-956 77492-358
(2-Bromo-6-fluoro-3-methoxyphenyl)(cyclopropyl)methanol ≥95%
(2-Bromo-6-fluoro-3-methoxyphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂BrFO₂
MW: 275.114 g/mol
Storage Temperature: Ambient
CAS Number: 3002475-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR