(2-Bromo-6-fluoro-3-methoxyphenyl)(4-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

323849-1G 323849-500MG 323849-5G
77469-276EA 415.09 USD
77469-276 77481-952 77492-354
(2-Bromo-6-fluoro-3-methoxyphenyl)(4-methoxyphenyl)methanol ≥95%
(2-Bromo-6-fluoro-3-methoxyphenyl)(4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄BrFO₃
Storage Temperature: Ambient
CAS Number: 3002513-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR