2-Chloro-2,2-difluoro-1-(perfluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

323798-1G 323798-500MG 323798-5G
77469-272EA 594.31 USD
77469-272 77481-948 77492-350
2-Chloro-2,2-difluoro-1-(perfluorophenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(perfluorophenyl)ethanone

Precaución: For research use only

Formula: C₈ClF₇O
MW: 280.527 g/mol
Storage Temperature: Ambient
CAS Number: 79542-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR