2-Ethyl-1-(perfluorophenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

323796-1G 323796-500MG 323796-5G
77469-268EA 415.09 USD
77469-268 77481-944 77492-346
2-Ethyl-1-(perfluorophenyl)butan-1-ol ≥95%
2-Ethyl-1-(perfluorophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃F₅O
MW: 268.223 g/mol
Storage Temperature: Ambient
CAS Number: 1713190-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR