propyl 3-hydroxy-4-methylhexanoate ≥95%

Supplier: AOBChem USA

Warning

323785-1G 323785-500MG 323785-5G
77469-256EA 415.09 USD
77469-256 77481-932 77492-334
propyl 3-hydroxy-4-methylhexanoate ≥95%
propyl 3-hydroxy-4-methylhexanoate

Precaución: For research use only

Formula: C₁₀H₂₀O₃
MW: 188.264 g/mol
Storage Temperature: Ambient
CAS Number: 3002508-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR