propyl 2-(1-hydroxycyclopentyl)acetate ≥95%

Supplier: AOBChem USA

Warning

323783-1G 323783-500MG 323783-5G
77469-252EA 347.11 USD
77469-252 77481-928 77492-330
propyl 2-(1-hydroxycyclopentyl)acetate ≥95%
propyl 2-(1-hydroxycyclopentyl)acetate

Precaución: For research use only

Formula: C₁₀H₁₈O₃
MW: 186.248 g/mol
Storage Temperature: Ambient
CAS Number: 1862971-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR