(5-Bromo-2-fluoro-3-methoxyphenyl)(furan-2-yl)methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
323720-1G
323720-500MG
323720-5G
77469-244EA
415.09
USD
77469-244
77481-920
77492-322
(5-Bromo-2-fluoro-3-methoxyphenyl)(furan-2-yl)methanol ≥95%
(5-Bromo-2-fluoro-3-methoxyphenyl)(furan-2-yl)methanol
Precaución: For research use only
Formula:
C₁₂H₁₀BrFO₃ MW: 301.108 g/mol Storage Temperature: Ambient |
CAS Number:
3002472-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...