(5-Bromo-2-fluoro-3-methoxyphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

323717-1G 323717-500MG 323717-5G
77469-240EA 415.09 USD
77469-240 77481-916 77492-318
(5-Bromo-2-fluoro-3-methoxyphenyl)(m-tolyl)methanol ≥95%
(5-Bromo-2-fluoro-3-methoxyphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄BrFO₂
Storage Temperature: Ambient
CAS Number: 3002479-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR