1-(5-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone 95

Supplier: AOBChem USA

Warning

323711-1G 323711-500MG 323711-5G
77469-236EA 415.09 USD
77469-236 77481-912 77492-314
1-(5-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone 95
1-(5-Bromo-2-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O₂
MW: 317.487 g/mol
Storage Temperature: Ambient
CAS Number: 3002458-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR