(5-Bromo-2-fluoro-3-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

323705-1G 323705-500MG 323705-5G
77469-232EA 415.09 USD
77469-232 77481-908 77492-310
(5-Bromo-2-fluoro-3-methoxyphenyl)(phenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-methoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₂BrFO₂
Storage Temperature: Ambient
CAS Number: 3002475-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR