(5-Fluoro-2-methoxyphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

323644-1G 323644-500MG 323644-5G
77469-228EA 415.09 USD
77469-228 77481-904 77492-306
(5-Fluoro-2-methoxyphenyl)(propyl)sulfane ≥95%
(5-Fluoro-2-methoxyphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃FOS
MW: 200.273 g/mol
Storage Temperature: Ambient
CAS Number: 3002421-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR