2,6-Dichloro-3-iodo-4-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

323528-1G 323528-500MG 323528-5G
77469-224EA 655.08 USD
77469-224 77481-900 77492-302
2,6-Dichloro-3-iodo-4-(trifluoromethyl)pyridine ≥95%
2,6-Dichloro-3-iodo-4-(trifluoromethyl)pyridine

Precaución: For research use only

Formula: C₆HCl₂F₃IN
MW: 341.885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827835
CAS Number: 1186405-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR