2,2-Difluoro-1-(5-methoxythiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

322815-1G 322815-500MG 322815-5G
77469-216EA 597.4 USD
77469-216 77481-892 77492-294
2,2-Difluoro-1-(5-methoxythiophen-2-yl)ethanone ≥95%
2,2-Difluoro-1-(5-methoxythiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₇H₆F₂O₂S
MW: 192.183 g/mol
Storage Temperature: Ambient
CAS Number: 1860279-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR