(4-pentylphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

322797-1G 322797-500MG 322797-5G
77469-212EA 415.09 USD
77469-212 77481-888 77492-290
(4-pentylphenyl)(m-tolyl)methanol ≥95%
(4-pentylphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₉H₂₄O
MW: 268.393 g/mol
Storage Temperature: Ambient
CAS Number: 3002536-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR