Isopropyl(4-pentylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

322790-1G 322790-500MG 322790-5G
77469-208EA 415.09 USD
77469-208 77481-884 77492-286
Isopropyl(4-pentylphenyl)sulfane ≥95%
Isopropyl(4-pentylphenyl)sulfane

Precaución: For research use only

Formula: C₁₄H₂₂S
Storage Temperature: Ambient
CAS Number: 2737205-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR