2-Fluoro-6-(methoxymethoxy)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32249-1G 32249-500MG 32249-5G
77469-204EA 544.87 USD
77469-204 77481-880 77492-282
2-Fluoro-6-(methoxymethoxy)phenylboronic acid pinacol ester ≥97%
2-Fluoro-6-(methoxymethoxy)phenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₄H₂₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD31619440
CAS Number: 1599432-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR