Ethyl 4-(4-fluorophenyl)-2-oxo-3-butynoate ≥95%

Supplier: AOBChem USA

Warning

321780-1G 321780-500MG 321780-5G
77469-200EA 192.61 USD
77469-200 77481-876 77492-276
Ethyl 4-(4-fluorophenyl)-2-oxo-3-butynoate ≥95%
Ethyl 4-(4-fluorophenyl)-2-oxo-3-butynoate

Precaución: For research use only

Formula: C₁₂H₉FO₃
Storage Temperature: Ambient
CAS Number: 1791418-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR