4-(4-Fluorophenyl)-3-butyn-2-one ≥95%

Supplier: AOBChem USA

Warning

321760-1G 321760-500MG 321760-5G
77469-196EA 364.62 USD
77469-196 77481-872 77492-272
4-(4-Fluorophenyl)-3-butyn-2-one ≥95%
4-(4-Fluorophenyl)-3-butyn-2-one

Precaución: For research use only

Formula: C₁₀H₇FO
MW: 162.16 g/mol
Storage Temperature: Ambient
CAS Number: 405065-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR