4-(4-Fluorophenyl)-2-methylbut-3-yn-2-ol ≥95%

Supplier: AOBChem USA

Warning

321749-1G 321749-500MG 321749-5G
77469-192EA 419.21 USD
77469-192 77481-868 77492-268
4-(4-Fluorophenyl)-2-methylbut-3-yn-2-ol ≥95%

Precaución: For research use only

Formula: C₁₁H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD01250505
CAS Number: 81233-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR