1-[2-(4-Methylphenyl)ethynyl]cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

321121-1G 321121-500MG 321121-5G
77469-188EA 364.62 USD
77469-188 77481-864 77492-264
1-[2-(4-Methylphenyl)ethynyl]cyclobutanol ≥95%
1-[2-(4-Methylphenyl)ethynyl]cyclobutanol

Precaución: For research use only

Formula: C₁₃H₁₄O
Storage Temperature: Ambient
CAS Number: 524742-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR