4-Methyl-1-(4-methylphenyl)-1-pentyn-3-ol ≥95%

Supplier: AOBChem USA

Warning

321109-1G 321109-500MG 321109-5G
77469-184EA 576.8 USD
77469-184 77481-860 77492-260
4-Methyl-1-(4-methylphenyl)-1-pentyn-3-ol ≥95%
4-Methyl-1-(4-methylphenyl)-1-pentyn-3-ol

Precaución: For research use only

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
Storage Temperature: Ambient
CAS Number: 288322-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR