4-(4-Methylphenyl)-3-butyn-2-ol ≥95%

Supplier: AOBChem USA

Warning

321087-1G 321087-500MG 321087-5G
77469-180EA 576.8 USD
77469-180 77481-856 77492-256
4-(4-Methylphenyl)-3-butyn-2-ol ≥95%
4-(4-Methylphenyl)-3-butyn-2-ol

Precaución: For research use only

Formula: C₁₁H₁₂O
Storage Temperature: Ambient
CAS Number: 79756-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR