1-(2,4-dimethoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

320467-1G 320467-500MG 320467-5G
77469-168EA 226.6 USD
77469-168 77481-844 77492-244
1-(2,4-dimethoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,4-dimethoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₃
MW: 224.296 g/mol
Storage Temperature: Ambient
CAS Number: 273411-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR