(2,6-Difluoro-4-methoxyphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

320335-1G 320335-500MG 320335-5G
77469-160EA 415.09 USD
77469-160 77481-836 77492-236
(2,6-Difluoro-4-methoxyphenyl)(propyl)sulfane ≥95%
(2,6-Difluoro-4-methoxyphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₂F₂OS
Storage Temperature: Ambient
CAS Number: 3002502-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR