2-(Benzyloxy)isonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

32013-1G 32013-500MG 32013-5G
77469-156EA 484.1 USD
77469-156 77481-832 77492-232
2-(Benzyloxy)isonicotinaldehyde ≥97%
2-(Benzyloxy)isonicotinaldehyde

Precaución: For research use only

Formula: C₁₃H₁₁NO₂
MW: 213.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256126
CAS Number: 467236-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR