Triethyl(4-propylphenyl)silane ≥95%
Supplier: AOBChem USA
|
Warning
|
319964-1G
319964-500MG
319964-5G
77469-152EA
415.09
USD
77469-152
77481-828
77492-226
Triethyl(4-propylphenyl)silane ≥95%
Triethyl(4-propylphenyl)silane
Precaución: For research use only
Formula:
C₁₅H₂₆Si MW: 234.452 g/mol Storage Temperature: Ambient |
CAS Number:
3002501-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...