2-(5-Bromo-2-fluoro-3-(methylthio)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

319913-1G 319913-500MG 319913-5G
77469-144EA 415.09 USD
77469-144 77481-820 77492-218
2-(5-Bromo-2-fluoro-3-(methylthio)phenyl)propan-2-ol ≥95%
2-(5-Bromo-2-fluoro-3-(methylthio)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFOS
MW: 279.169 g/mol
Storage Temperature: Ambient
CAS Number: 3002532-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR