(2-(Benzyloxy)-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

319909-1G 319909-500MG 319909-5G
77469-140EA 415.09 USD
77469-140 77481-816 77492-214
(2-(Benzyloxy)-5-methylphenyl)trimethylsilane ≥95%
(2-(Benzyloxy)-5-methylphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₇H₂₂OSi
MW: 270.441 g/mol
Storage Temperature: Ambient
CAS Number: 3002425-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR