1-(2-(Benzyloxy)-5-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

319907-1G 319907-500MG 319907-5G
77469-136EA 415.09 USD
77469-136 77481-812 77492-210
1-(2-(Benzyloxy)-5-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-(Benzyloxy)-5-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₉H₂₄O₂
Storage Temperature: Ambient
CAS Number: 3002470-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR