(2-(Benzyloxy)-5-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

319905-1G 319905-500MG 319905-5G
77469-132EA 415.09 USD
77469-132 77481-808 77492-206
(2-(Benzyloxy)-5-methylphenyl)(cyclopropyl)methanol ≥95%
(2-(Benzyloxy)-5-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₈H₂₀O₂
Storage Temperature: Ambient
CAS Number: 3002453-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR