(2-(Benzyloxy)-5-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

319901-1G 319901-500MG 319901-5G
77469-128EA 415.09 USD
77469-128 77481-804 77492-202
(2-(Benzyloxy)-5-methylphenyl)(ethyl)sulfane ≥95%
(2-(Benzyloxy)-5-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈OS
MW: 258.379 g/mol
Storage Temperature: Ambient
CAS Number: 3002485-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR