1-(2-(Benzyloxy)-5-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

319897-1G 319897-500MG 319897-5G
77469-124EA 415.09 USD
77469-124 77481-800 77492-198
1-(2-(Benzyloxy)-5-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(2-(Benzyloxy)-5-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₇H₁₃F₅O₂
MW: 344.276 g/mol
Storage Temperature: Ambient
CAS Number: 3002435-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR