2-(Ethylthio)dibenzo[b,d]furan ≥95%

Supplier: AOBChem USA

Warning

319828-1G 319828-500MG 319828-5G
77469-120EA 415.09 USD
77469-120 77481-796 77492-194
2-(Ethylthio)dibenzo[b,d]furan ≥95%
2-(Ethylthio)dibenzo[b,d]furan

Precaución: For research use only

Formula: C₁₄H₁₂OS
MW: 228.3094 g/mol
Storage Temperature: Ambient
CAS Number: 3002475-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR