2-Chloro-1-(dibenzo[b,d]furan-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

319825-1G 319825-500MG 319825-5G
77469-116EA 415.09 USD
77469-116 77481-792 77492-190
2-Chloro-1-(dibenzo[b,d]furan-2-yl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(dibenzo[b,d]furan-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₄H₇ClF₂O₂
MW: 280.6539 g/mol
Storage Temperature: Ambient
CAS Number: 3002454-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR