2-(3-(Benzyloxy)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

319821-1G 319821-500MG 319821-5G
77469-112EA 96.82 USD
77469-112 77481-788 77492-186
2-(3-(Benzyloxy)phenyl)propan-2-ol ≥95%
2-(3-(Benzyloxy)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
CAS Number: 79678-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR