(3-(Benzyloxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

319817-1G 319817-500MG 319817-5G
77469-108EA 415.09 USD
77469-108 77481-784 77492-182
(3-(Benzyloxy)phenyl)(methyl)sulfane ≥95%
(3-(Benzyloxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₄OS
MW: 230.3253 g/mol
Storage Temperature: Ambient
CAS Number: 1450897-58-1
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR