1-(2-Methyl-4-((4-Methylbenzyl)oxy)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

319813-1G 319813-500MG 319813-5G
77469-104EA 415.09 USD
77469-104 77481-780 77492-178
1-(2-Methyl-4-((4-Methylbenzyl)oxy)phenyl)propan-1-one ≥95%
1-(2-Methyl-4-((4-Methylbenzyl)oxy)phenyl)propan-1-one

Precaución: For research use only

Formula: C₁₈H₂₀O₂
MW: 268.3502 g/mol
Storage Temperature: Ambient
CAS Number: 3002484-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR