Ethyl(2-methyl-4-((4-methylbenzyl)oxy)phenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

319805-1G 319805-500MG 319805-5G
77469-100EA 415.09 USD
77469-100 77481-776 77492-174
Ethyl(2-methyl-4-((4-methylbenzyl)oxy)phenyl)sulfane ≥95%
Ethyl(2-methyl-4-((4-methylbenzyl)oxy)phenyl)sulfane

Precaución: For research use only

Formula: C₁₇H₂₀OS
Storage Temperature: Ambient
CAS Number: 3002435-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR