Isopropyl(2-methyl-4-((4-methylbenzyl)oxy)phenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

319803-1G 319803-500MG 319803-5G
77469-096EA 415.09 USD
77469-096 77481-772 77492-170
Isopropyl(2-methyl-4-((4-methylbenzyl)oxy)phenyl)sulfane ≥95%
Isopropyl(2-methyl-4-((4-methylbenzyl)oxy)phenyl)sulfane

Precaución: For research use only

Formula: C₁₈H₂₂OS
Storage Temperature: Ambient
CAS Number: 3002475-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR