2,2-Difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

319800-1G 319800-500MG 319800-5G
77469-092EA 415.09 USD
77469-092 77481-768 77492-166
2,2-Difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone ≥95%
2,2-Difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₆F₂O₂
MW: 290.3045 g/mol
Storage Temperature: Ambient
CAS Number: 3002483-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR