2-(4-((4-methoxybenzyl)oxy)-2-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

319794-1G 319794-500MG 319794-5G
77469-088EA 415.09 USD
77469-088 77481-764 77492-162
2-(4-((4-methoxybenzyl)oxy)-2-methylphenyl)propan-2-ol ≥95%
2-(4-((4-methoxybenzyl)oxy)-2-methylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₈H₂₂O₃
MW: 286.3654 g/mol
Storage Temperature: Ambient
CAS Number: 3002484-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR