Ethyl(4-((4-methoxybenzyl)oxy)-2-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

319789-1G 319789-500MG 319789-5G
77469-084EA 415.09 USD
77469-084 77481-760 77492-158
Ethyl(4-((4-methoxybenzyl)oxy)-2-methylphenyl)sulfane ≥95%
Ethyl(4-((4-methoxybenzyl)oxy)-2-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₇H₂₀O₂S
Storage Temperature: Ambient
CAS Number: 3002435-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR