2,2-Difluoro-1-(4-((4-methoxybenzyl)oxy)-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

319784-1G 319784-500MG 319784-5G
77469-080EA 415.09 USD
77469-080 77481-756 77492-154
2,2-Difluoro-1-(4-((4-methoxybenzyl)oxy)-2-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-((4-methoxybenzyl)oxy)-2-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₆F₂O₃
MW: 306.3039 g/mol
Storage Temperature: Ambient
CAS Number: 3002453-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR