1-(Benzo[d][1,3]dioxol-5-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

319702-1G 319702-500MG 319702-5G
77469-076EA 538.69 USD
77469-076 77481-752 77492-150
1-(Benzo[d][1,3]dioxol-5-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-(Benzo[d][1,3]dioxol-5-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅ClF₂O₃
MW: 234.5840 g/mol
Storage Temperature: Ambient
CAS Number: 1338328-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR