1-([1,1'-biphenyl]-3-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

319698-1G 319698-500MG 319698-5G
77469-072EA 415.09 USD
77469-072 77481-748 77492-146
1-([1,1'-biphenyl]-3-yl)cyclobutanol ≥95%
1-([1,1'-biphenyl]-3-yl)cyclobutanol

Precaución: For research use only

Formula: C₁₆H₁₆O
Storage Temperature: Ambient
CAS Number: 2790604-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR