[1,1'-biphenyl]-3-yl(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

319696-1G 319696-500MG 319696-5G
77469-068EA 415.09 USD
77469-068 77481-744 77492-142
[1,1'-biphenyl]-3-yl(phenyl)methanol ≥95%
[1,1'-biphenyl]-3-yl(phenyl)methanol

Precaución: For research use only

Formula: C₁₉H₁₆O
MW: 260.3297 g/mol
Storage Temperature: Ambient
CAS Number: 873384-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR