1-(2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

318605-1G 318605-500MG 318605-5G
77469-056EA 477.92 USD
77469-056 77481-732 77492-126
1-(2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)-2,2,2-trifluoroethanone ≥95%
1-(2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₈H₅F₃O₃S
MW: 238.1837 g/mol
Storage Temperature: Ambient
CAS Number: 2028004-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR